Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium chromate, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Cesium chromate
CAS 13454-78-9
Formula Weight 381.81
MDL Number MFCD00016050
UN Number UN3087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1349214

  SDS
alfa aesar™
1349214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1349222

  SDS
alfa aesar™
1349222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 381.81
Chemical Name or Material Cesium chromate
TSCA TSCA
Odor Odorless
Physical Form Powder
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00016050
CAS 13454-78-9
UN Number UN3087
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.