Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium chromate, 99+%

Chemical Identifiers

Chemical Name or Material Cesium chromate
CAS 13454-78-9
Molecular Formula Cs2CrO4
Formula Weight 381.81
MDL Number MFCD00016050
UN Number UN3087
Synonym Chromic acid cesium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2361909

  SDS
alfa aesar™
B2361909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2361918

  SDS
alfa aesar™
B2361918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2361930

  SDS
alfa aesar™
B2361930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 381.81
Health Hazard 2 GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
TSCA TSCA
MDL Number MFCD00016050
Molecular Formula Cs2CrO4
Synonym Chromic acid cesium salt
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
Chemical Name or Material Cesium chromate
Quantity 10g
Health Hazard 1 Danger
Assay ≥99%
Odor Odorless
CAS 13454-78-9
UN Number UN3087
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.