missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium chloride, White powder/crystalline/beads, 99.99% (Metals basis), Alfa Aesar™

$181.61 - $2,307.95

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.355
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4372009
View Documents
Thermo Fisher Scientific
4372009
10g Each for $181.61
 
AA4372018
View Documents
Thermo Fisher Scientific
4372018
50g Each for $559.31
 
AA4372030
View Documents
Thermo Fisher Scientific
4372030
250g Each for $2,307.95
 
Specifications

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
CHEBI:63039
[Cl-].[Cs+]

Specifications

7647-17-8
ClCs
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
3.99
1.6418
1303°C
646°C
Cesium chloride
99.99% (Metals basis)
MFCD00010955
Soluble in water and alcohol. Insoluble in acetone.
[Cl-].[Cs+]
168.355
CHEBI:63039
-10 Mesh Beads,Ampouled under argon
Odorless
Hygroscopic
White
10g
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P308+P313-P501c

H361-H373

EINECSNumber : 231-600-2

RTECSNumber : FK9625000

TSCA : Yes

Recommended Storage : Ambient temperatures