missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer:  Avantor Performance 404202

 View more versions of this product

Catalog No. 02002569



Description

Description

    For Density Gradient Centrifugation; For Laboratory,Research,or Manufacturing Use
Specifications

Specifications

Cesium Chloride
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
Poly Bottle
1290°C
1kg
7647-17-8
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
[Cl-].[Cs+]
168.355
CHEBI:63039
Ultrapure Bioreagent
1L = 3.99kg
646°C

Chemical Identifiers

7647-17-8
168.355
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
Documents