missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium chloride, ultrapure, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ J6595014

 View more versions of this product

Catalog No. AAJ6595014



Specifications

Specifications

Cesium chloride
Ultrapure
ClCs
14,2011
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
99.9%
Odorless
Hygroscopic
White
25g
7647-17-8
(metals basis)
MFCD00010955
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
[Cl-].[Cs+]
168.355
CHEBI:63039
Crystalline
3.99
1.6418
1303°C
646°C

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
CHEBI:63039
[Cl-].[Cs+]
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P308+P313-P501c

H361-H373

EINECSNumber : 231-600-2

RTECSNumber : FK9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents