missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium chloride, Puriss. p.a., ≥99.5%, Honeywell Fluka™
Puriss. p.a.,≥99.5%
$880.13 - $4677.43
Chemical Identifiers
| CAS | 7647-17-8 |
|---|---|
| Molecular Formula | ClCs |
| Molecular Weight (g/mol) | 168.36 |
| MDL Number | MFCD00010955 |
| InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
| Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
| PubChem CID | 24293 |
| ChEBI | CHEBI:63039 |
| IUPAC Name | cesium;chloride |
| SMILES | [Cl-].[Cs+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-17-8 | |
| 168.36 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| 24293 | |
| cesium;chloride |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| CHEBI:63039 | |
| [Cl-].[Cs+] |
Specifications
| 7647-17-8 | |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| [Cl-].[Cs+] | |
| 168.36 | |
| CHEBI:63039 | |
| ≥99.5% | |
| Poly bottle |
| 100 g | |
| CsCl | |
| NONH for all modes of transport | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| cesium;chloride | |
| 24293 | |
| 168.36g/mol | |
| puriss. p.a. | |
| Cesium Chloride |
Safety and Handling
P280
H361
EINECSNumber : 231-600-2
RTECSNumber : FK9625000