missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride, For Analysis, EMSURE™, MilliporeSigma™

$211.51 - $634.74

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.355
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1020380025
View Documents
Emd Millipore Corporation
1020380025
25g Glass bottle Each for $211.51
 
M1020380100
View Documents
Emd Millipore Corporation
1020380100
100g Plastic bottle Each for $634.74
 
Description

Description

For analysis
Specifications

Chemical Identifiers

7647-17-8
168.355
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
CHEBI:63039
[Cl-].[Cs+]
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride

Specifications

7647-17-8
White solid
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
Odorless
25g
ClCs
caesium 1+ ion chloride, caesium chloride, cesium chloride, cesium chloride cscl, cesium monochloride, cscl, dicesium dichloride, gnr9hml8ba, tricesium trichloride, unii-gnr9hml8ba
[Cl-].[Cs+]
168.355
CHEBI:63039
Solid
Glass bottle
Cesium chloride