Please login to your online account to display your discounted pricing

Cesium Chloride (Crystalline Powder), Fisher BioReagents


For isolation of phage DNA, plasmid DNA, or total RNA

Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP1595500

 View more versions of this product

Catalog No. BP1595-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Physical Form Powder/Solid
Formula Weight 168.36
Boiling Point 1290°C
Melting Point 646°C
Quantity 500g
Merck Index 15, 2010
Molecular Formula ClCs
pH 5 to 9
Color White
DNase DNase free
Protease Protease free
Potassium (K) 0.03% max.
Assay Percent Range ≥99.5 %
ChemAlert Storage Symbol Gray
CAS 7647-17-8
Chemical Name or Material Cesium Chloride
Lithium (Li) 0.0002% max.
Packaging Poly Bottle
Sodium (Na) 0.02% max.
Refractive Index 1.364 to 1.374 (at 25°C)
Absorbance 0.08 max. (50% w/v solution) at 260nm
Purity Grade Notes DNase-, RNase- and Protease-Free
Recommended Storage RT

Specifications

Chemical Name or Material Cesium Chloride
CAS 7647-17-8
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Powder/Solid
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula ClCs
Formula Weight 168.36
Accessories
 

RT

SDS & Safety Documents