missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, BiotechGrade, 99.9%, Spectrum™ Chemical

C1252, 7647-17-8, CsCl

379,73 $ - 1 157,70 $

Identifiants chimiques

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
18601545
Afficher les documents
Spectrum Chemical Mfg Cor
C125225GM
25 g Poly Bottle N/A
Il en reste null
 
18601546
Afficher les documents
Spectrum Chemical Mfg Cor
C1252100GM
100 g Poly Bottle N/A
Il en reste null
 
Description

Description

Spectrum™ Chemical Cesium Chloride, BiotechGrade is a colorless inorganic compound and a solid. It is used in isopynic centrifugation as well as in analytical chemistry as a reagent.
Spécifications

Identifiants chimiques

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
[Cl-].[Cs+]
ClCs
MFCD00010955
caesium(1+) chloride

Spécifications

Fine, white, crystal
100%
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
99.9%
Poly Bottle
7647-17-8
ClCs
25 g
[Cl-].[Cs+]
168.36
Biotech
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'