missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium chloride, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1288618

 View more versions of this product

Catalog No. AA1288618



Description

Description

  • Evacuation of radio tubes, manufacturing of cesium compounds, brewing, x-ray fluorescent screens, radiographic contrast medium
Specifications

Specifications

Cesium chloride
99% (Metals basis)
ClCs
14,2011
Very soluble in water; Soluble in alcohol; Insoluble in acetone
[Cl-].[Cs+]
168.355
CHEBI:63039
Crystalline
3.99g/mL
1.6418
1303°C
50g
7647-17-8
(metals basis)
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
99%
Odorless
Hygroscopic
646°C

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
Safety and Handling

Safety and Handling

H303 : May be harmful if swallowed.

RTECSNumber : FK9625000

TSCA : TSCA

Documents