Learn More
Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 189502500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1290.0°C | |
646.0°C | |
7647-17-8 | |
Crystalline Powder | |
99% min. (Argentometry) | |
CsCl | |
15, 2010 | |
Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
≥99% | |
Plastic bottle |
Cesium chloride | |
p.a. | |
White | |
250 g | |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
caesium(1+) chloride | |
24293 | |
168.36 | |
Analytical | |
3.983 |
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
[Cl-].[Cs+] |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 231-600-2
RTECSNumber : FK9625000
TSCA : TSCA
RUO – Research Use Only