missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride, 99.999% (Metals Basis), Molecular Biology Grade, Ultrapure, Affymetrix/USB™

Used for density gradient high speed centrifugation separation of λ-phage, M13 and plasmid DNA.

Manufacturer:  Thermo Fisher Scientific J7582222

 View more versions of this product

Catalog No. AAJ7582222



Specifications

Specifications

Cesium Chloride
Metal basis
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
99.999%
Plastic bottle
1303°C
100g
7647-17-8
ClCs
caesium 1+ ion chloride
[Cl-].[Cs+]
168.355
CHEBI:63039
Crystalline Powder
Molecular Biology
3.99
646°C

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
Safety and Handling

Safety and Handling

TSCA : Y

Documents