missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium chloride, 99.999+% (metals basis)

Manufacturer:  Alfa Aesar™ 8918818

 View more versions of this product

Catalog No. AA8918818



Specifications

Specifications

Cesium chloride
(metals basis)
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
[Cl-].[Cs+]
168.355
CHEBI:63039
Powder
3.99g/mL
1.6418
1303°C
50g
7647-17-8
ClCs
14,2011
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
≥99.999%
Odorless
Hygroscopic
646°C

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
Safety and Handling

Safety and Handling

RTECSNumber : FK9625000

TSCA : TSCA

SDS Documents