missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium chloride, 99.999+% (metals basis)

Manufacturer:  Alfa Aesar™ 8918809

 View more versions of this product

Catalog No. AA8918809



Specifications

Specifications

Cesium chloride
≥99.999%
ClCs
14,2011
[Cl-].[Cs+]
168.355
CHEBI:63039
Powder
Odorless
Hygroscopic
646°C
7647-17-8
(metals basis)
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
3.99g/mL
1.6418
1303°C
10g

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
CHEBI:63039
[Cl-].[Cs+]
ClCs
MFCD00010955
24293
cesium;chloride
Safety and Handling

Safety and Handling

RTECSNumber : FK9625000

TSCA : TSCA

SDS Documents