missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 206325000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysis, For UV/VIS spectroscopySpecifications
Cesium chloride | |
7647-17-8 | |
99.99% | |
CsCl | |
15, 2010 | |
Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol,insoluble in acetone | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
99.99% | |
Glass bottle |
For spectroscopy | |
500g | |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
cesium;chloride | |
24293 | |
168.36 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 231-600-2
RUO â Research Use Only