missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade, ACROS Organics™

Manufacturer:  Acros Organics AC206320050

 View more versions of this product

Catalog No. AC206320050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

For ultra trace analysis, For UV/VIS spectroscopy
Specifications

Specifications

(50% aqueous solution) 1 cm layer
For spectroscopy
(50%, aqua soln)
ClCs
MFCD00010955
caesium 1+ ion chloride
[Cl-].[Cs+]
168.355
CHEBI:63039
Liquid
Trace Metal Basis
Glass bottle
Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
Colorless to Yellow
5g
Cesium chloride
7647-17-8
99.99%
CsCl
15, 2010
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
99.99%
25ppm max.
99.94% min. (metal based, calc.)
1290.0°C
646.0°C

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
SDS Documents