missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium chloride, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8764009

 View more versions of this product

Catalog No. AA8764009



Specifications

Specifications

Cesium chloride
99.99%
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
[Cl-].[Cs+]
168.355
CHEBI:63039
Crystalline
Odorless
10g
7647-17-8
ClCs
White powder/crystalline/beads
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
3.99g/cm3 at 20°C
White

Chemical Identifiers

7647-17-8
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
Safety and Handling

Safety and Handling

H303 : May be harmful if swallowed.

EINECSNumber : 231-600-2

SDS Documents