Learn More
Cesium chloride, 99.9% (metals basis), Thermo Scientific Chemicals
$325.45 - $325.45
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
PubChem CID | 24293 |
ChEBI | CHEBI:63039 |
IUPAC Name | caesium(1+) chloride |
SMILES | [Cl-].[Cs+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1001822
|
Thermo Scientific Chemicals
01001822 |
100 g |
Each for $325.45
|
|
|||||
Description
Analytical chemistry reagent for detecting inorganic ions. Source of cesium metal. Phase transfer catalyst in organic chemistry. Used for density gradient work to separate and purify proteins (isopycnic centrifugation) including chromosomal, plasmid and organelle (mitochondrial or plasmid) DNAs, and different RNA types. Preparation of electrically conducting glasses and screens of cathode ray tubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
caesium(1+) chloride |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 | |
[Cl-].[Cs+] |
Specifications
1303°C | |
7647-17-8 | |
100 g | |
ClCs | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
caesium(1+) chloride | |
24293 | |
168.36 | |
Hygroscopic | |
Cesium chloride |
646°C | |
Crystalline | |
99.9% (metals basis) | |
MFCD00010955 | |
Soluble in water and concentrated hydrochloric acid. | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
1.6418 | |
3.99 |
Safety and Handling
P201-P202-P260-P281-P308+P313-P501c
H361-H373
EINECSNumber : 231-600-2
RTECSNumber : FK9625000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only