Please login to your online account to display your discounted pricing

Cesium carbonate, 99% (metals basis), Alfa Aesar™

Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes

Chemical Identifiers

Chemical Name or Material Cesium carbonate
CAS 534-17-8
Molecular Formula Cs2CO3
Formula Weight 325.82
MDL Number MFCD00010957
Beilstein 4546405
Merck Index 14,2010
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA12887A1

  SDS
alfa aesar
12887A1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 325.82
Quantity 1kg
Chemical Name or Material Cesium carbonate
TSCA TSCA
Odor Odorless
Physical Form Powder/Granules
Assay 99%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Molecular Formula Cs2CO3
MDL Number MFCD00010957
CAS 534-17-8
Beilstein 4546405
Merck Index 14,2010
Color White
Melting Point 610°C (dec.)
pH 10 to 12
Solubility Information Very soluble in water and alcohol. Soluble in ether
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.072
RTECS Number FK9400000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.