missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium carbonate, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8763509

 View more versions of this product

Catalog No. AA8763509



Specifications

Specifications

Cesium carbonate
99.99% (Metal basis)
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
C(=O)([O-])[O-].[Cs+].[Cs+]
325.819
325.82
4.072g/cm3 at 20°C
White
534-17-8
CCs2O3
White powder
FJDQFPXHSGXQBY-UHFFFAOYSA-L
dicesium;carbonate
10796
Crystalline
Odorless
10g

Chemical Identifiers

534-17-8
325.819
FJDQFPXHSGXQBY-UHFFFAOYSA-L
10796
C(=O)([O-])[O-].[Cs+].[Cs+]
CCs2O3
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
dicesium;carbonate
Safety and Handling

Safety and Handling

H315-H319-H335-H303 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

EINECSNumber : 208-591-9

SDS Documents