Please login to your online account to display your discounted pricing

Cesium carbonate, 99.9% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cesium carbonate
CAS 534-17-8
Name Note (metals basis)
Molecular Formula Cs2CO3
Formula Weight 325.82
MDL Number MFCD00010957
Beilstein 4546405
Merck Index 14,2010
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1092436

 
alfa aesar
1092436
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 325.82
Quantity 500g
Chemical Name or Material Cesium carbonate
TSCA TSCA
Mesh Size −20 Mesh
Odor Odorless
Name Note (metals basis)
Physical Form Powder/Granules
Assay 99.9%
Sensitivity Hygroscopic
Molecular Formula Cs2CO3
MDL Number MFCD00010957
CAS 534-17-8
Beilstein 4546405
Merck Index 14,2010
Melting Point 610°C (Dec.)
pH 10 to 12
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.072
RTECS Number FK9400000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.