missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium bromide, White powder/crystalline, 99% (Metals basis), Alfa Aesar™

$83.73 - $281.26

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.809
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym br.cs, caesium 1+ ion bromide, caesium bromide, caesium i bromide, cesium bromide, cesium bromide csbr, cesiumbromide, csbr, tricesium tribromide, unii-06m25edm3f
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1349018
View Documents
Thermo Fisher Scientific
1349018
50g Each for $83.73
 
AA1349030
View Documents
Thermo Fisher Scientific
1349030
250g Each for $281.26
 
Specifications

Chemical Identifiers

7787-69-1
212.809
LYQFWZFBNBDLEO-UHFFFAOYSA-M
24592
[Br-].[Cs+]
BrCs
MFCD00010954
br.cs, caesium 1+ ion bromide, caesium bromide, caesium i bromide, cesium bromide, cesium bromide csbr, cesiumbromide, csbr, tricesium tribromide, unii-06m25edm3f
cesium;bromide

Specifications

7787-69-1
BrCs
br.cs, caesium 1+ ion bromide, caesium bromide, caesium i bromide, cesium bromide, cesium bromide csbr, cesiumbromide, csbr, tricesium tribromide, unii-06m25edm3f
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium;bromide
24592
Crystalline
Odorless
Hygroscopic
White
50g
99% (Metals basis)
MFCD00010954
Very soluble in water. Soluble in alcohol. Insoluble in acetone
[Br-].[Cs+]
212.809
212.81
4.44 g/cm3 at 20°C
1.6984
1300°C
636°C
Cesium bromide
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H361

EINECSNumber : 232-130-0

RTECSNumber : FK9275000

TSCA : Yes

Recommended Storage : Ambient temperatures