missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium bromide, White powder/crystalline, 99.9% (Metals basis), Alfa Aesar™

$117.18 - $459.66

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.809
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym br.cs, caesium 1+ ion bromide, caesium bromide, caesium i bromide, cesium bromide, cesium bromide csbr, cesiumbromide, csbr, tricesium tribromide, unii-06m25edm3f
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1292818
View Documents
Thermo Fisher Scientific
1292818
50g Each for $117.18
 
AA1292830
View Documents
Thermo Fisher Scientific
1292830
250g Each for $459.66
 
Specifications

Chemical Identifiers

7787-69-1
212.809
LYQFWZFBNBDLEO-UHFFFAOYSA-M
24592
[Br-].[Cs+]
BrCs
MFCD00010954
br.cs, caesium 1+ ion bromide, caesium bromide, caesium i bromide, cesium bromide, cesium bromide csbr, cesiumbromide, csbr, tricesium tribromide, unii-06m25edm3f
cesium;bromide

Specifications

7787-69-1
BrCs
br.cs, caesium 1+ ion bromide, caesium bromide, caesium i bromide, cesium bromide, cesium bromide csbr, cesiumbromide, csbr, tricesium tribromide, unii-06m25edm3f
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium;bromide
24592
Crystalline
Odorless
Hygroscopic
White
50g
99.9% (Metals basis)
MFCD00010954
Soluble in water.
[Br-].[Cs+]
212.809
212.81
4.44
1.6984
1300°C
636°C
Cesium bromide
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H361

EINECSNumber : 232-130-0

RTECSNumber : FK9275000

TSCA : Yes

Recommended Storage : Ambient temperatures