missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium bromide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 8763909

 View more versions of this product

Catalog No. AA8763909



Specifications

Specifications

Cesium bromide
99.999%
BrCs
MFCD00010954
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium;bromide
24592
4.44g/mL
Odorless
Hygroscopic
White
10g
7787-69-1
(metals basis)
CsBr
14,2009
[Br-].[Cs+]
212.809
212.81
−4 Mesh
1.6984
1300°C
636°C

Chemical Identifiers

7787-69-1
212.809
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium;bromide
BrCs
MFCD00010954
24592
[Br-].[Cs+]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FK9275000

TSCA : TSCA

SDS Documents