missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium bromide, 99.9%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 192060250

 View more versions of this product

Catalog No. AC192060250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1300.0°C
636.0°C
7787-69-1
Crystalline Powder and/or Crystals
99.90%
CsBr
15, 2008
Solubility in water: 55g/100mL (20 c). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
[Br-].[Cs+]
212.81
212.81
Pure
4.44
Cesium bromide
99.9%
White
25 g
BrCs
MFCD00010954
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium;bromide
24592
99.9%
Glass bottle
4.4400g/mL

Chemical Identifiers

7787-69-1
212.81
LYQFWZFBNBDLEO-UHFFFAOYSA-M
24592
[Br-].[Cs+]
BrCs
MFCD00010954
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
cesium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 232-130-0

RTECSNumber : FK9275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only