Learn More
Cesium bromide, 99.9%, pure, Thermo Scientific Chemicals
$151.18 - $151.18
Chemical Identifiers
CAS | 7787-69-1 |
---|---|
Molecular Formula | BrCs |
Molecular Weight (g/mol) | 212.81 |
MDL Number | MFCD00010954 |
InChI Key | LYQFWZFBNBDLEO-UHFFFAOYSA-M |
Synonym | cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide |
PubChem CID | 24592 |
IUPAC Name | cesium;bromide |
SMILES | [Br-].[Cs+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC192060250
|
Thermo Scientific Chemicals
192060250 |
25 g | Glass bottle |
Each for $151.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-69-1 | |
212.81 | |
LYQFWZFBNBDLEO-UHFFFAOYSA-M | |
24592 | |
[Br-].[Cs+] |
BrCs | |
MFCD00010954 | |
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide | |
cesium;bromide |
Specifications
1300.0°C | |
7787-69-1 | |
Crystalline Powder and/or Crystals | |
99.90% | |
CsBr | |
15, 2008 | |
Solubility in water: 55g/100mL (20 c). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone | |
[Br-].[Cs+] | |
212.81 | |
212.81 | |
Pure | |
4.44 | |
Cesium bromide, 99.9% |
636.0°C | |
White | |
25 g | |
BrCs | |
MFCD00010954 | |
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide | |
LYQFWZFBNBDLEO-UHFFFAOYSA-M | |
cesium;bromide | |
24592 | |
99.9% | |
Glass bottle | |
4.4400g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
EINECSNumber : 232-130-0
RTECSNumber : FK9275000
TSCA : TSCA
RUO – Research Use Only