missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium acetate, 99%, for analysis, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC390830250

 View more versions of this product

Catalog No. AC390830250



Description

Description

Specifications

Specifications

Cesium acetate
0.5ppm max.
C2H3CsO2
MFCD00013056
Solubility in water: 91.5% (21°C)
CC(=O)[O-].[Cs+]
10ppm max.
5152919
Crystalline Powder, Crystals, and/or Chunks
Analysis
50ppm max.
Authentic
1ppm max.
0.5ppm max.
5ppm max.
Solubility in water: 91.5% (21°C).
White
25g
3396-11-0
99%
CH3CO2Cs
acetic acid cesium, acetic acid cesium salt, acetic acid, cesium salt, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, caesium 1+ acetate ion, caesium 1+ ion acetate, caesium acetate, cesium acetate, cesiumacetate
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
cesium;acetate
191.96
191.96
99%
5ppm max.
0.5ppm max.
0.5ppm max.
1ppm max.
Glass bottle
10ppm max.
5ppm max.
194.0°C

Chemical Identifiers

3396-11-0
191.96
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
5152919
CC(=O)[O-].[Cs+]
C2H3CsO2
MFCD00013056
acetic acid cesium, acetic acid cesium salt, acetic acid, cesium salt, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, caesium 1+ acetate ion, caesium 1+ ion acetate, caesium acetate, cesium acetate, cesiumacetate
cesium;acetate
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.

GHS Signal Word: Warning

EINECSNumber : 222-248-

SDS Documents