missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cesium acetate, 99%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 191041000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cesium acetate | |
3396-11-0 | |
Glass bottle | |
CH3CO2Cs | |
100g | |
Solubility in water: 91.5% (21°C). | |
CC(=O)[O-].[Cs+] | |
191.96 | |
191.96 | |
Pure |
Pure | |
99% | |
C2H3CsO2 | |
MFCD00013056 | |
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
cesium;acetate | |
5152919 | |
99% |
Chemical Identifiers
3396-11-0 | |
191.96 | |
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
5152919 | |
CC(=O)[O-].[Cs+] |
C2H3CsO2 | |
MFCD00013056 | |
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
cesium;acetate |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.
GHS Signal Word: Warning
EINECSNumber : 222-248-
RUO â Research Use Only