Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium acetate, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Cesium acetate
CAS 3396-11-0
Formula Weight 191.95
MDL Number MFCD00013056
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1292914

 
alfa aesar™
1292914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1292922

 
alfa aesar™
1292922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.95
Chemical Name or Material Cesium acetate
TSCA TSCA
Odor Weak
Physical Form Crystalline
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00013056
CAS 3396-11-0
Melting Point 194°C
Quantity 25g