missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium acetate, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1292922

 View more versions of this product

Catalog No. AA1292922



Specifications

Specifications

Cesium acetate
(metals basis)
MFCD00013056
Soluble in water,DMSO.
CC(=O)[O-].[Cs+]
191.949
191.95
99.9%
Hygroscopic
100g
3396-11-0
C2H3CsO2
acetic acid cesium
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
cesium;acetate
5152919
Crystalline
Weak
194°C

Chemical Identifiers

3396-11-0
191.949
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
5152919
CC(=O)[O-].[Cs+]
C2H3CsO2
MFCD00013056
acetic acid cesium
cesium;acetate
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361

EINECSNumber : 222-248-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents