missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium AA Standard, 1mL = 1mg Cs (1,000ppm Cs), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIACS1KN500

 View more versions of this product

Catalog No. ACS1KN-500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Cesium AA Standard
94.0,4.22,0.14
Liquid
1,000ppm Cs
Clear colorless
7732-18-5,7697-37-2,7789-18-6
97.0,4.39,0.14
Poly natural
1 mL = 1 mg Cs
500mL

Chemical Identifiers

7732-18-5,7697-37-2,7789-18-6
97.0,4.39,0.14
Cesium AA Standard
94.0,4.22,0.14
Liquid