missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cesium AA Standard, 1000 ppm Cs in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company ACS1KN100
Specifications
Cesium AA Standard | |
Natural LDPE Bottle | |
94.0,4.22,0.14 | |
Colorless | |
CsNO3 | |
[N+](=O)([O-])[O-].[Cs+] | |
194.909 | |
1 mL = 1 mg Cs |
7789-18-6,7697-37-2,7789-18-6 | |
100 mL | |
97.0,4.39,0.14 | |
1,000ppm Cs | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
cesium;nitrate | |
62674 |
Chemical Identifiers
7789-18-6 | |
194.909 | |
62674 | |
[N+](=O)([O-])[O-].[Cs+] |
CsNO3 | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
cesium;nitrate |