Please login to your online account to display your discounted pricing

Cesium, 99.98% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cesium
CAS 7440-46-2
Name Note (metals basis)
MDL Number MFCD00134037
UN Number UN1407
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1014614

 
alfa aesar
1014614
Vacuum sealed in breakseal ampoules 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Cesium
TSCA TSCA
Name Note (metals basis)
Assay 99.98%
MDL Number MFCD00134037
CAS 7440-46-2
UN Number UN1407
Odor Odorless
Packaging Vacuum sealed in breakseal ampoules
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Golden
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: CAESIUM
RTECS Number FK9225000
Safety and Handling

Danger

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.