missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium, 99.98% (metals basis), Alfa Aesar

Manufacturer:  Alfa Aesar 1014603

 View more versions of this product

Catalog No. AA1014603



Specifications

Specifications

Cesium
7440-46-2
99.98% (Metals basis)
MFCD00134037
TVFDJXOCXUVLDH-UHFFFAOYSA-N
cesium
132.905
CHEBI:30514
Solid
Silver
1g
Silver-colored solid
Cs
1ksv9v4y4i
[Cs]
Odorless
5354618
132.9054
0.0000011 hPa

Chemical Identifiers

7440-46-2
132.905
TVFDJXOCXUVLDH-UHFFFAOYSA-N
5354618
cesium
Cs
MFCD00134037
1ksv9v4y4i
CHEBI:30514
[Cs]
Safety and Handling

Safety and Handling

H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-155-4

Documents