missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Verispec™ Certified Reference KCl Conductivity Standard
Certified reference material/standard. KCl conductivity standard.
$179.72 - $179.72
Specifications
Chemical Name or Material | KCl |
---|---|
Technique | Calibration of Conductivity Meter |
Color | Colorless |
For Use With (Equipment) | Conductivity Meters and Probes |
Solubility | Soluble |
Catalog Number | Mfr. No. | Conductivity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conductivity | Price | Quantity | |||||
01-185-100
|
Verispec™
RV588860500A |
25,000μS/cm at 25°C |
Each for $179.72
|
|
|||||
01-185-101
|
Verispec™
RV588843500A |
5000μS/cm at 25°C |
Each for $179.72
|
|
|||||
01-185-102
|
Verispec™
RV588775500A |
750μS/cm at 25°C |
Each for $179.72
|
|
|||||
01-185-103
|
Verispec™
RV588750500A |
500μS/cm at 25°C |
Each for $179.72
|
|
|||||
01-185-104
|
Verispec™
RV588684500A |
84μS/cm at 25°C |
Each for $179.72
|
|
|||||
Description
- Verispec™ Certified Reference Material/Standard
- KCl Conductivity Standard
- Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Specifications
KCl | |
Colorless | |
Soluble | |
Certified Reference Material |
Calibration of Conductivity Meter | |
Conductivity Meters and Probes | |
Conductivity Standard | |
Certified |