Learn More
Cerium zirconium oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03921636
Description
Used as wear resistant grinding and dispersion media. Used in fuel cell materials (SOFC), body armors. Also finds applications in catalytic converter, honey comb, precision ceramic parts, abrasive wheels, oxygen sensors, cutting tools. Used as ceramic steel. Other uses include screen printing, dip and spin coating and APS coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium zirconium oxide | |
Powder | |
(metals basis) | |
Cerium zirconate | |
295.32 |
53169-24-7 | |
500 g | |
MFCD00168089 | |
Soluble in acids. Insoluble in water. | |
99.5% |
Chemical Identifiers
53169-24-7 | |
MFCD00168089 | |
Cerium zirconate |
Safety and Handling
EINECSNumber : 258-407-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only