Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium vanadium oxide, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Cerium vanadium oxide
CAS 13597-19-8
Formula Weight 255.06g/mol
MDL Number MFCD00171262
UN Number UN3285
Synonym Cerium vanadate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4020914

 
alfa aesar™
40209-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4020922

 
alfa aesar™
40209-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Mesh Size −200 Mesh
Quantity 25g
Quantity 25g
Chemical Name or Material Cerium vanadium oxide
CAS 13597-19-8
Physical Form Powder
Assay 99.9%
Assay Percent Notes (REO)
Formula Weight 255.06g/mol
MDL Number MFCD00171262
UN Number UN3285
Synonym Cerium vanadate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.