Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium tungsten oxide, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Cerium tungsten oxide
CAS 13454-74-5
Formula Weight 1023.78
MDL Number MFCD00049725
Synonym Cerium tungstate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3584014

 
alfa aesar™
3584014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3584022

 
alfa aesar™
3584022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1023.78
Chemical Name or Material Cerium tungsten oxide
TSCA TSCA
Mesh Size −100 Mesh
Odor Odorless
Physical Form Powder
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00049725
CAS 13454-74-5
Synonym Cerium tungstate
Melting Point 1089°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.