missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium sputtering target, 76.2mm dia x 3.18mm thick

Chemical Identifiers

CAS 7440-45-1
MDL Number MFCD00010924
UN Number UN1333
Chemical Name or Material Cerium sputtering target, 76.2mm dia. x 3.18mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45926KS SDS Alfa Aesar™
45926KS
1 ea. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7440-45-1
MFCD00010924
UN1333
Cerium sputtering target, 76.2mm dia. x 3.18mm thick

Specifications

1 ea.
Odorless
UN1333
7440-45-1
MFCD00010924
Cerium sputtering target, 76.2mm dia. x 3.18mm thick

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

TSCA : TSCA