Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium sputtering target, 50.8mm dia x 1.59mm thick

Chemical Identifiers

Chemical Name or Material Cerium sputtering target
CAS 7440-45-1
Name Note 50.8mm dia. x 1.59mm thick
MDL Number MFCD00010924
UN Number UN1333
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45618KS

 
alfa aesar™
45618KS
1 ea. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Cerium sputtering target
Odor Odorless
TSCA TSCA
Name Note 50.8mm dia. x 1.59mm thick
MDL Number MFCD00010924
CAS 7440-45-1
UN Number UN1333
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM
Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.