missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cerium silicide, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03583606
Description
Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium silicide | |
Gray | |
Pieces | |
(REO) | |
MFCD00168082 | |
Insoluble in water. | |
[Si]=[Ce]=[Si] | |
196.286 | |
196.29 | |
Odorless |
1620°C | |
12014-85-6 | |
5 g | |
CeSi2 | |
cerium silicide, cerium disilicide | |
BLHYVSKDMXEWHD-UHFFFAOYSA-N | |
bis($l^{2}-silanylidene)cerium | |
6336874 | |
99.9% | |
5.31 g/mL |
Chemical Identifiers
12014-85-6 | |
196.286 | |
BLHYVSKDMXEWHD-UHFFFAOYSA-N | |
6336874 | |
[Si]=[Ce]=[Si] |
CeSi2 | |
MFCD00168082 | |
cerium silicide, cerium disilicide | |
bis($l^{2}-silanylidene)cerium |
Safety and Handling
EINECSNumber : 234-602-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only