missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium rod, 6.35mm (0.25 in.) dia., 99.9% (metals basis excluding Ta)

Manufacturer:  Alfa Aesar™ 44037HF

 View more versions of this product

Catalog No. AA44037HF



Description

Description

  • Packaged in mylar under argon
Specifications

Specifications

Cerium rod
10 mm
7440-45-1
Ce
UN1333
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerium
140.116
23974
≈0.21g/mm,Cast Rod,diameter is approximate
Silver
≈0.21g/mm
≈6.35mm (0.25 in.) dia.
(metals basis excluding Ta)
MFCD00010924
acmc-1bl5r
[Ce]
Odorless
Mylar under Argon
CHEBI:33369
99.9%

Chemical Identifiers

7440-45-1
140.116
GWXLDORMOJMVQZ-UHFFFAOYSA-N
23974
cerium
Ce
MFCD00010924
acmc-1bl5r
CHEBI:33369
[Ce]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

EINECSNumber : 231-154-9

TSCA : Yes

storageNote1 :

  • Packaged in mylar under argon

Recommended Storage : Ambient temperatures

Documents