Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta)

$578.59 - $578.59

Chemical Identifiers

Chemical Name or Material Cerium rod
CAS 7440-45-1
Name Note 12.7mm (0.5 in.) dia.
MDL Number MFCD00010924
UN Number UN1333
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40284DH

  SDS
alfa aesar™
40284DH
25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40284DJ

  SDS
alfa aesar™
40284DJ
50mm Each for $578.59

AA40284DM

  SDS
alfa aesar™
40284DM
100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Cast Rod, dia. is approximate
Quantity 25mm
Packaging Mylar under Argon
Name Note 12.7mm (0.5 in.) dia.
Assay Percent Notes (metals basis excluding Ta)
CAS 7440-45-1
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM
Weight ≈0.86g/mm
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Color Silver
Assay 99.9%
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
UN Number UN1333
Chemical Name or Material Cerium rod
MDL Number MFCD00010924
Storage Note 1
  • Packaged under argon
Odor Odorless

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

  • Packaged under argon