missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium, plasma standard solution, Ce 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13815AE

 View more versions of this product

Catalog No. AA13815AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cerium
Clear liquid
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerium
23974
Liquid
Colorless
7440-45-1,7440-45-1,7732-18-5
Ce
Nitric acid: 5.0%; Cerium: 0.1%; Water: 94.9%
[Ce]
140.116
CHEBI:33369
23 hPa (17mm Hg) at 20°C
100mL

Chemical Identifiers

7440-45-1
140.116
GWXLDORMOJMVQZ-UHFFFAOYSA-N
CHEBI:33369
[Ce]
Ce
MFCD00010924
23974
cerium
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents