missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cerium, plasma standard solution, Ce 1000μg/mL, Thermo Scientific Chemicals, Specpure™
Supplier: Thermo Scientific Chemicals 013815AD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium | |
50 mL | |
Ce | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
cerium | |
23974 | |
Nitric acid: 5.0%; Cerium: 0.1%; Water: 94.9% | |
Liquid |
7440-45-1,7732-18-5 | |
Colorless | |
MFCD00010924 | |
[Ce] | |
140.12 | |
CHEBI:33369 | |
23 hPa (17mm Hg) at 20°C |
Chemical Identifiers
7440-45-1 | |
140.12 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
CHEBI:33369 | |
[Ce] |
Ce | |
MFCD00010924 | |
23974 | |
cerium |
Safety and Handling
P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
H290-H315-H319
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only