missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cerium, plasma standard solution, Ce 1000μg/mL, Alfa Aesar™, Specpure™
Manufacturer: Alfa Aesar 13815AP
Specifications
Cerium | |
Clear liquid | |
MFCD00010924 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
cerium | |
23974 | |
Liquid | |
Colorless |
7440-45-1,7440-45-1,7732-18-5 | |
Ce | |
Nitric acid: 5.0%; Cerium: 0.1%; Water: 94.9% | |
[Ce] | |
140.116 | |
CHEBI:33369 | |
23 hPa (17mm Hg) at 20°C | |
500mL |
Chemical Identifiers
7440-45-1 | |
140.116 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
CHEBI:33369 | |
[Ce] |
Ce | |
MFCD00010924 | |
23974 | |
cerium |
Safety and Handling
H314: Causes severe skin burns and eye damage.
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-714-2