missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L2005614

 View more versions of this product

Catalog No. AAL2005614



Description

Description

Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
Specifications

Specifications

Cerium(IV) trifluoromethanesulfonate
C4CeF12O12S4
25 g
cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
BYAJTISDHIASIP-UHFFFAOYSA-J
cerium(4+);trifluoromethanesulfonate
9875257
98%
107792-63-2
MFCD03844746
Hygroscopic
Soluble in water.
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
736.37
736.39

Chemical Identifiers

107792-63-2
736.37
BYAJTISDHIASIP-UHFFFAOYSA-J
9875257
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
C4CeF12O12S4
MFCD03844746
cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
cerium(4+);trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H500

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only