missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) sulfate tetrahydrate, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 199130025

Catalog No. AC199130025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cerium(IV) sulfate tetrahydrate
10294-42-5
98% min. (Iodometry)
Ce(SO4)2·4H2O
02,65; 15,73
cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate
MEXSQFDSPVYJOM-UHFFFAOYSA-J
cerium(4+);disulfate;tetrahydrate
22723557
98+%
1.06
99+%
2.5kg
CeO8S2·4H2O
MFCD00149427
14, 1990
Solubility in water: 38g/L (50°C). Other solubilities: soluble in dilute sulfuric acid,slowly soluble in cold,more readily soluble in,hot mineral acids
O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
404.29
404.29
Plastic bottle
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only