missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) sulfate hydrate, REacton™, 99% (REO)

Manufacturer:  Alfa Aesar™ 1132336

 View more versions of this product

Catalog No. AA1132336



Specifications

Specifications

Cerium(IV) sulfate hydrate
(REO)
MFCD00149427
acmc-20mqih, cerium 4+ sulfate-water 1/2/1, cerium iv sulfate hydrate, reacton, cerium iv sulfate monohydrate, sulfuricacid, cerium salt, hydrate 9ci
KNANFSXYDCURQS-UHFFFAOYSA-J
cerium(4+);disulfate;hydrate
20223355
Crystalline
Reacton™
Moisture sensitive
500g
95838-16-7
CeH2O9S2
UN3085
Slightly soluble in water. Soluble in dil.sulfuric acid.
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
350.243
332.24 (Anhydrous)
99%
3.01 g/mL
195°C (decomposition)

Chemical Identifiers

95838-16-7
350.243
KNANFSXYDCURQS-UHFFFAOYSA-J
20223355
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
CeH2O9S2
MFCD00149427
acmc-20mqih, cerium 4+ sulfate-water 1/2/1, cerium iv sulfate hydrate, reacton, cerium iv sulfate monohydrate, sulfuricacid, cerium salt, hydrate 9ci
cerium(4+);disulfate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

EINECSNumber : 237-029-5

RTECSNumber : WS6960000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents