Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) sulfate hydrate, 98%

$55.88 - $55.88

Chemical Identifiers

Chemical Name or Material Cerium (IV) sulfate hydrate
CAS 95838-16-7
Molecular Formula Ce(SO4)2·xH2O
Formula Weight 332.24 (Anhydrous)
MDL Number MFCD00149427
UN Number UN3085
Merck Index 14,1990
Synonym Ceric sulfate hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1014218

 
alfa aesar™
A1014218
50g Each for $55.88

AAA101420B

 
alfa aesar™
A101420B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1014230

 
alfa aesar™
A1014230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 332.24 (Anhydrous)
Health Hazard 3 GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
CAS 95838-16-7
UN Number UN3085
Merck Index 14,1990
Melting Point 195°C (dec.)
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
RTECS Number WS6960000
Chemical Name or Material Cerium (IV) sulfate hydrate
Assay 98%
Molecular Formula Ce(SO4)2·xH2O
MDL Number MFCD00149427
Synonym Ceric sulfate hydrate
Quantity 50g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.

Applications
Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed

Safety and Handling

Danger

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.